3,4-Difluoro-L-phenylalanine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(S)-2-Amino-3-(3,4-difluorophenyl)propanoic acid, H-Phe(3,4-DiF)-OH
PC8363-25G
PC8363-5G
APOSPC8363-25GEA
21230
CZK
APOSPC8363-25G
APOSPC8363-5G
3,4-Difluoro-L-phenylalanine
3,4-Difluoro-L-phenylalanine
Vzorec:
C₉H₉F₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01075125 CAS číslo: 31105-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...