Fmoc-2-(trifluoromethyl)-L-phenylalanine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Fmoc-Phe(2-CF₃)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid, 2-(Trifluoromethyl)-L-phenylalanine, N-FMOC protected
PC8356-5G
APOSPC8356-5GEA
6520
CZK
APOSPC8356-5G
Fmoc-2-(trifluoromethyl)-L-phenylalanine
Fmoc-2-(trifluoromethyl)-L-phenylalanine
Vzorec:
C₂₅H₂₀F₃NO₄ MW: 455,44 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01863052 CAS číslo: 352523-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...