1,4-Dibromo-1,1,2-trifluorobutane 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC8340-25G
PC8340-5G
APOSPC8340-25GEA
17390
CZK
APOSPC8340-25G
APOSPC8340-5G
1,4-Dibromo-1,1,2-trifluorobutane 95%
1,4-Dibromo-1,1,2-trifluorobutane
Vzorec:
C₄H₅Br₂F₃ Storage Temperature: Ambient |
MDL Number:
MFCD04038280 CAS číslo: 155957-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...