Tisk…

3-Hydroxy-5-(trifluoromethyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Hydroxy-5-(trifluoromethyl)phenyl)boronic acid

PC8280-1G PC8280-250MG
APOSPC8280-1GEA 8630 CZK
APOSPC8280-1G APOSPC8280-250MG
3-Hydroxy-5-(trifluoromethyl)phenylboronic acid
3-Hydroxy-5-(trifluoromethyl)phenylboronic acid
Vzorec: C₇H₆BF₃O₃
MW: 205,930 g/mol
Bod tání: 92…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878337
CAS číslo: 957062-66-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR