1H,1H-Heptafluorobutyl acetate 97%
Dodavatel: Apollo Scientific
Synonyms:
2,2,3,3,4,4,4-heptafluorobutyl acetate
PC8275-25G
PC8275-5G
PC8275-100G
APOSPC8275-25GEA
5000
CZK
APOSPC8275-25G
APOSPC8275-5G
APOSPC8275-100G
1H,1H-Heptafluorobutyl acetate 97%
1H,1H-Heptafluorobutyl acetate
Vzorec:
C₆H₅F₇O₂ MW: 242,09 g/mol Bod varu: 105 °C/735mmHg Hustota: 1,435 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01070838 CAS číslo: 356-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...