Tisk…

4-(Trifluoromethylthio)phenylacetic acid 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid

PC8253-25G PC8253-5G PC8253-1G
APOSPC8253-25GEA 8700 CZK
APOSPC8253-25G APOSPC8253-5G APOSPC8253-1G
4-(Trifluoromethylthio)phenylacetic acid 96%
4-(Trifluoromethylthio)phenylacetic acid
Vzorec: C₉H₇F₃O₂S
MW: 236,208 g/mol
Bod tání: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00236351
CAS číslo: 243977-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR