4-(2-Fluoro-4-nitrophenyl)morpholine
Dodavatel: Apollo Scientific
PC8240-1G
PC8240-5G
APOSPC8240-1GEA
2070
CZK
APOSPC8240-1G
APOSPC8240-5G
4-(2-Fluoro-4-nitrophenyl)morpholine
4-(2-Fluoro-4-nitrophenyl)morpholine
Vzorec:
C₁₀H₁₁FN₂O₃ MW: 226,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03138381 CAS číslo: 2689-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...