Tisk…

4-Bromo-2,6-difluorobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Bromo-2,6-difluorophenyl)methanol

PC8239-1G PC8239-25G
APOSPC8239-1GEA 0 CZK
APOSPC8239-1G APOSPC8239-25G
4-Bromo-2,6-difluorobenzyl alcohol
4-Bromo-2,6-difluorobenzyl alcohol
Vzorec: C₇H₅BrF₂O
MW: 223,02 g/mol
Bod tání: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03094461
CAS číslo: 162744-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR