Tisk…

6-Fluoro-4-quinolinol 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Fluoro-4-hydroxyquinoline

PC8235-1G PC8235-5G PC8235-10G
APOSPC8235-1GEA 705 CZK
APOSPC8235-1G APOSPC8235-5G APOSPC8235-10G
6-Fluoro-4-quinolinol 98%
6-Fluoro-4-quinolinol
Vzorec: C₉H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD00269610
CAS číslo: 391-78-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR