[4-(Trifluoromethyl)phenoxy]acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(4-(Trifluoromethyl)phenoxy)acetic acid
PC8233-1G
PC8233-250MG
PC8233-5G
PC8233-10G
APOSPC8233-1GEA
1950
CZK
APOSPC8233-1G
APOSPC8233-250MG
APOSPC8233-5G
APOSPC8233-10G
[4-(Trifluoromethyl)phenoxy]acetic acid
[4-(Trifluoromethyl)phenoxy]acetic acid
Vzorec:
C₉H₇F₃O₃ MW: 220,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094458 CAS číslo: 163839-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...