Tisk…

4-Chloro-3-(trifluoromethyl)benzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC8231-5G PC8231-1G
APOSPC8231-5GEA 787 CZK
APOSPC8231-5G APOSPC8231-1G
4-Chloro-3-(trifluoromethyl)benzylamine 98%
4-Chloro-3-(trifluoromethyl)benzylamine
Vzorec: C₈H₇ClF₃N
Bod varu: 100…104 °C/12mmHg
Hustota: 1,373 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00190124
CAS číslo: 62039-92-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR