Tisk…

3-Fluoro-p-cresol

Dodavatel: Apollo Scientific
Nebezpečí

PC8221-1G PC8221-25G PC8221-5G
APOSPC8221-1GEA 622 CZK
APOSPC8221-1G APOSPC8221-25G APOSPC8221-5G
3-Fluoro-p-cresol
3-Fluoro-p-cresol
Vzorec: C₇H₇FO
Storage Temperature: Ambient
MDL Number: MFCD00143164
CAS číslo: 452-78-8
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR