Tisk…

4-Chloro-3-fluorobenzyl alcohol 96%

Dodavatel: Apollo Scientific
Varování

PC8146-1G PC8146-25G PC8146-5G
APOSPC8146-1GEA 622 CZK
APOSPC8146-1G APOSPC8146-25G APOSPC8146-5G
4-Chloro-3-fluorobenzyl alcohol 96%
4-Chloro-3-fluorobenzyl alcohol
Vzorec: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD00143292
CAS číslo: 202925-10-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR