3-Bromo-N-cyclopropyl-5-(trifluoromethyl)benzenesulphonamide
Dodavatel: Apollo Scientific
Synonyms:
3-Bromo-5-(N-cyclopropylsulphamoyl)benzotrifluoride
PC8108-1G
PC8108-250MG
APOSPC8108-1GEA
8100
CZK
APOSPC8108-1G
APOSPC8108-250MG
3-Bromo-N-cyclopropyl-5-(trifluoromethyl)benzenesulphonamide
3-Bromo-N-cyclopropyl-5-(trifluoromethyl)benzenesulphonamide
Vzorec:
C₁₀H₉BrF₃NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD09801015 CAS číslo: 951884-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...