Tisk…

Methyl-4-fluoro-2-nitrobenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 4-fluoro-2-nitrobenzoate, 4-Fluorobenzoic acid methyl ester

PC8106-1G PC8106-25G PC8106-5G
APOSPC8106-1GEA 992 CZK
APOSPC8106-1G APOSPC8106-25G APOSPC8106-5G
Methyl-4-fluoro-2-nitrobenzoate
Methyl-4-fluoro-2-nitrobenzoate
Vzorec: C₈H₆FNO₄
MW: 199.14
Storage Temperature: Ambient
MDL Number: MFCD09800921
CAS číslo: 151504-81-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR