Tisk…

2'-Chloro-4',5'-difluoroacetophenone 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(2-Chloro-4,5-difluorophenyl)ethanone, 1-chloro-3,4-difluor-6-ethanoylbenzene

PC8096-10G PC8096-1G
APOSPC8096-10GEA 9530 CZK
APOSPC8096-10G APOSPC8096-1G
2'-Chloro-4',5'-difluoroacetophenone 95%
2'-Chloro-4',5'-difluoroacetophenone
Vzorec: C₈H₅ClF₂O
MW: 190.58
Storage Temperature: Ambient
MDL Number: MFCD00671761
CAS číslo: 121872-94-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR