3-Bromo-5-iodobenzotrifluoride 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-5-iodo-alpha,alpha,alpha-trifluorotoluene, 1-Bromo-3-iodo-5-(trifluoromethyl)benzene
PC8091-1G
PC8091-5G
PC8091-10G
APOSPC8091-1GEA
622
CZK
APOSPC8091-1G
APOSPC8091-5G
APOSPC8091-10G
3-Bromo-5-iodobenzotrifluoride 98%
3-Bromo-5-iodobenzotrifluoride
Vzorec:
C₇H₃BrF₃I Storage Temperature: Refrigerator |
MDL Number:
MFCD09033749 CAS číslo: 481075-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...