2-Bromo-N-cyclohexyl-5-fluorobenzamide
Dodavatel: Apollo Scientific
PC8082-1G
PC8082-5G
APOSPC8082-1GEA
3340
CZK
APOSPC8082-1G
APOSPC8082-5G
2-Bromo-N-cyclohexyl-5-fluorobenzamide
2-Bromo-N-cyclohexyl-5-fluorobenzamide
Vzorec:
C₁₃H₁₅BrFNO Storage Temperature: Ambient |
MDL Number:
MFCD09800915 CAS číslo: 951884-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...