5-Methoxy-2-(trifluoromethyl)pyridine 98+%
Dodavatel: Apollo Scientific
|
Varování
|
PC8070-1G
PC8070-25G
PC8070-5G
APOSPC8070-1GEA
1040
CZK
APOSPC8070-1G
APOSPC8070-25G
APOSPC8070-5G
5-Methoxy-2-(trifluoromethyl)pyridine 98+%
5-Methoxy-2-(trifluoromethyl)pyridine
Vzorec:
C₇H₆F₃NO MW: 177,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09864699 CAS číslo: 216766-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...