Tisk…

Xenon difluoride 99,9%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Xenon(II) fluoride

PC8020-1G PC8020-25G PC8020-5G
APOSPC8020-1GEA 2490 CZK
APOSPC8020-1G APOSPC8020-25G APOSPC8020-5G
Xenon difluoride 99,9%
Xenon difluoride

Convenient fluorinating agent

Vzorec: F₂Xe
MW: 169.29
Bod varu: 114
Bod tání: 129 °C
Hustota: 4.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040538
CAS číslo: 13709-36-9
EINECS: 237-251-2
UN: 3087
ADR: 5.1+6.1 II
Merck Index: 14,10072

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR