Tisk…

2,6-Difluorophenylboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,6-Difluorobenzeneboronic acid pinacol ester, 2,6-Difluorophenylboronic acid, pinacol ester, 2-(2,6-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC8017-1G PC8017-5G
APOSPC8017-1GEA 622 CZK
APOSPC8017-1G APOSPC8017-5G
2,6-Difluorophenylboronic acid pinacol ester 98%
2,6-Difluorophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₅BF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09801043
CAS číslo: 863868-37-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR