2-Nitro-4-(trifluoromethyl)phenylacetonitrile 98%
Dodavatel: Apollo Scientific
Synonyms:
2-(2-Nitro-4-(trifluoromethyl)phenyl)acetonitrile
PC8009-10G
PC8009-1G
PC8009-5G
APOSPC8009-10GEA
4470
CZK
APOSPC8009-10G
APOSPC8009-1G
APOSPC8009-5G
2-Nitro-4-(trifluoromethyl)phenylacetonitrile 98%
2-Nitro-4-(trifluoromethyl)phenylacetonitrile
Vzorec:
C₉H₅F₃N₂O₂ MW: 230,15 g/mol Bod varu: 134…138 °C Bod tání: 38…34 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045621 CAS číslo: 13544-06-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...