Tisk…

3-Chloro-5-(trifluoromethyl)anisole 98%

Dodavatel: Apollo Scientific
Varování

PC8005-5G PC8005-1G
APOSPC8005-5GEA 4860 CZK
APOSPC8005-5G APOSPC8005-1G
3-Chloro-5-(trifluoromethyl)anisole 98%
3-Chloro-5-(trifluoromethyl)anisole
Vzorec: C₈H₆ClF₃O
MW: 210,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115855
CAS číslo: 886497-07-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR