Tisk…

4-Amino-3-fluorothiophenol

Dodavatel: Apollo Scientific
Varování

PC7991-1G PC7991-5G
APOSPC7991-1GEA 3310 CZK
APOSPC7991-1G APOSPC7991-5G
4-Amino-3-fluorothiophenol
4-Amino-3-fluorothiophenol
Vzorec: C₆H₆FNS
Storage Temperature: Refrigerator
MDL Number: MFCD09835280
CAS číslo: 15178-48-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR