Tisk…

Vanadium(IV) fluoride 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Vanadium tetrafluoride

PC7980-100G PC7980-25G
APOSPC7980-100GEA 8570 CZK
APOSPC7980-100G APOSPC7980-25G
Vanadium(IV) fluoride 95%
Vanadium(IV) fluoride
Vzorec: F₄V
MW: 126,94 g/mol
Bod tání: 325 °C
Hustota: 2,975 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042554
CAS číslo: 10049-16-8
EINECS: 233-171-7
UN: 3285
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR