3-Fluoro-4-hydroxybenzoic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC7964-100G
PC7964-25G
PC7964-5G
APOSPC7964-100GEA
6880
CZK
APOSPC7964-100G
APOSPC7964-25G
APOSPC7964-5G
3-Fluoro-4-hydroxybenzoic acid
3-Fluoro-4-hydroxybenzoic acid
Vzorec:
C₇H₅FO₃ MW: 156,11 g/mol Bod tání: 161…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143161 CAS číslo: 350-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...