Tisk…

Ultramark®

Dodavatel: Apollo Scientific
Varování

Synonyms: SpectraSynQ PFPE-1600

PC7953-100MG PC7953-1G
APOSPC7953-100MGEA 622 CZK
APOSPC7953-100MG APOSPC7953-1G
Ultramark®
Ultramark®

Fluorinated polyether for mass spectrometry up to mass 1960

Bod varu: 128 °C/0.2mmHg
Storage Temperature: Ambient
MDL Number: MFCD00166131
CAS číslo: 66795-94-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR