3-Bromo-2-fluoropyridine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC7951-5G
PC7951-100G
PC7951-25G
APOSPC7951-5GEA
622
CZK
APOSPC7951-5G
APOSPC7951-100G
APOSPC7951-25G
3-Bromo-2-fluoropyridine 98%
3-Bromo-2-fluoropyridine
Vzorec:
C₅H₃BrFN Bod varu: 76 °C/20mmHg Bod vzplanutí: <90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04112496 CAS číslo: 36178-05-9 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...