2-Iodo-6-nitro-4-(trifluoromethyl)aniline 95+%
Dodavatel: Apollo Scientific
PC7943-1G
PC7943-500MG
PC7943-5G
APOSPC7943-1GEA
7670
CZK
APOSPC7943-1G
APOSPC7943-500MG
APOSPC7943-5G
2-Iodo-6-nitro-4-(trifluoromethyl)aniline 95+%
2-Iodo-6-nitro-4-(trifluoromethyl)aniline
Vzorec:
C₇H₄F₃IN₂O₂ Bod tání: 102…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD14560546 CAS číslo: 543740-74-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...