Tisk…

4-Fluoro-o-anisidine hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-2-methoxyaniline hydrochloride

PC7914-1G PC7914-25G PC7914-5G
APOSPC7914-1GEA 622 CZK
APOSPC7914-1G APOSPC7914-25G APOSPC7914-5G
4-Fluoro-o-anisidine hydrochloride 98%
4-Fluoro-o-anisidine hydrochloride
Vzorec: C₇H₉ClFNO
MW: 177,6000 g/mol
Bod tání: 201…209 °C
Storage Temperature: Ambient
MDL Number: MFCD09800850
CAS číslo: 178671-97-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR