2,4-Difluoro-3-methylbenzonitrile 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC7906-1G
PC7906-5G
APOSPC7906-1GEA
992
CZK
APOSPC7906-1G
APOSPC7906-5G
2,4-Difluoro-3-methylbenzonitrile 99%
2,4-Difluoro-3-methylbenzonitrile
Vzorec:
C₈H₅F₂N MW: 153,1288 g/mol Bod tání: 42…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD07777155 CAS číslo: 847502-87-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...