Tisk…

2-Fluoro-4-methoxybenzyl alcohol

Dodavatel: Apollo Scientific
Varování

PC7889-1G PC7889-5G
APOSPC7889-1GEA 4440 CZK
APOSPC7889-1G APOSPC7889-5G
2-Fluoro-4-methoxybenzyl alcohol
2-Fluoro-4-methoxybenzyl alcohol
Vzorec: C₈H₉FO₂
MW: 156,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116336
CAS číslo: 405-09-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR