5-Fluoro-2-hydroxy-N,N-dimethylbenzylamine 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-[(Dimethylamino)methyl]-4-fluorophenol
PC7876-1G
PC7876-5G
APOSPC7876-1GEA
1620
CZK
APOSPC7876-1G
APOSPC7876-5G
5-Fluoro-2-hydroxy-N,N-dimethylbenzylamine 99%
5-Fluoro-2-hydroxy-N,N-dimethylbenzylamine
Vzorec:
C₉H₁₂FNO MW: 169,20 g/mol Bod varu: 225 °C Hustota: 1,14 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03412238 CAS číslo: 46049-91-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...