4,5-Difluoro-2-methylbenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC7873-1G
PC7873-250MG
PC7873-5G
APOSPC7873-1GEA
2070
CZK
APOSPC7873-1G
APOSPC7873-250MG
APOSPC7873-5G
4,5-Difluoro-2-methylbenzoic acid 98%
4,5-Difluoro-2-methylbenzoic acid
Vzorec:
C₈H₆F₂O₂ MW: 172,13 g/mol Bod tání: 117…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD08062377 CAS číslo: 183237-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...