2,3-Difluoro-4-methoxybenzoic acid 98%
Dodavatel: Apollo Scientific
PC7872-10G
PC7872-5G
PC7872-1G
APOSPC7872-10GEA
4470
CZK
APOSPC7872-10G
APOSPC7872-5G
APOSPC7872-1G
2,3-Difluoro-4-methoxybenzoic acid 98%
2,3-Difluoro-4-methoxybenzoic acid
Vzorec:
C₈H₆F₂O₃ Bod tání: 222…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD03428569 CAS číslo: 329014-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...