Tisk…

5'-Fluoro-2'-(trifluoromethyl)acetophenone 97%

Dodavatel: Apollo Scientific

PC7863-10G PC7863-5G PC7863-1G
APOSPC7863-10GEA 6750 CZK
APOSPC7863-10G APOSPC7863-5G APOSPC7863-1G
5'-Fluoro-2'-(trifluoromethyl)acetophenone 97%
5'-Fluoro-2'-(trifluoromethyl)acetophenone
Vzorec: C₉H₆F₄O
MW: 206.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236274
CAS číslo: 243863-39-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR