3,4-Difluorophenylglycine
Dodavatel: Apollo Scientific
PC7856-1G
PC7856-5G
PC7856-10G
APOSPC7856-1GEA
4530
CZK
APOSPC7856-1G
APOSPC7856-5G
APOSPC7856-10G
3,4-Difluorophenylglycine
3,4-Difluorophenylglycine
Vzorec:
C₈H₇F₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06246973 CAS číslo: 225641-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...