2-Mercapto-4-(trifluoromethyl)pyrimidine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Sulphanyl-4-(trifluoromethyl)pyrimidine
PC7832-1G
PC7832-5G
PC7832-250MG
APOSPC7832-1GEA
2700
CZK
APOSPC7832-1G
APOSPC7832-5G
APOSPC7832-250MG
2-Mercapto-4-(trifluoromethyl)pyrimidine 97%
2-Mercapto-4-(trifluoromethyl)pyrimidine
Vzorec:
C₅H₃F₃N₂S MW: 180,148 g/mol Bod tání: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00115113 CAS číslo: 136547-17-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...