Tisk…

2,4,6-Trifluorophenol 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC7817P-25G PC7817P-5G PC7817P-1G
APOSPC7817P-25GEA 9660 CZK
APOSPC7817P-25G APOSPC7817P-5G APOSPC7817P-1G
2,4,6-Trifluorophenol 98%
2,4,6-Trifluorophenol
Vzorec: C₆H₃F₃O
MW: 148,08 g/mol
Bod tání: 47…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00061216
CAS číslo: 2268-17-9
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR