4-Chloro-2-fluorobenzoyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC7798-25G
PC7798-5G
PC7798-1G
APOSPC7798-25GEA
6390
CZK
APOSPC7798-25G
APOSPC7798-5G
APOSPC7798-1G
4-Chloro-2-fluorobenzoyl chloride
4-Chloro-2-fluorobenzoyl chloride
Vzorec:
C₇H₃Cl₂FO Bod varu: 105…106 °C/14mmHg Bod vzplanutí: 78 °C Storage Temperature: Ambient |
CAS číslo:
394-39-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...