1-(Trifluoromethyl)vinyl acetate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,3,3-Trifluoroprop-1-en-2-yl acetate
PC7782-100G
PC7782-25G
PC7782-5G
APOSPC7782-100GEA
14490
CZK
APOSPC7782-100G
APOSPC7782-25G
APOSPC7782-5G
1-(Trifluoromethyl)vinyl acetate 97%
1-(Trifluoromethyl)vinyl acetate
Vzorec:
C₅H₅F₃O₂ MW: 154,09 g/mol Bod varu: 86…88 °C Hustota: 1,2 g/cm³ (25 °C) Bod vzplanutí: 2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040844 CAS číslo: 2247-91-8 EINECS: 218-838-2 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...