1,3-Dichloro-2,4-difluoro-5-nitrobenzene 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC7781-1G
PC7781-5G
APOSPC7781-1GEA
6450
CZK
APOSPC7781-1G
APOSPC7781-5G
1,3-Dichloro-2,4-difluoro-5-nitrobenzene 95+%
1,3-Dichloro-2,4-difluoro-5-nitrobenzene
Vzorec:
C₆HCl₂F₂NO₂ Storage Temperature: Ambient |
CAS číslo:
15952-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...