5-(Trifluoromethyl)uracil 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Trifluorothymine, 5-(Trifluoromethyl)pyrimidine-2,4(1H,3H)-dione
PC7780-25G
PC7780-100G
APOSPC7780-25GEA
7410
CZK
APOSPC7780-25G
APOSPC7780-100G
5-(Trifluoromethyl)uracil 98%
5-(Trifluoromethyl)uracil
Vzorec:
C₅H₃F₃N₂O₂ MW: 180,09 g/mol Bod tání: 245…249 °C Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006024 CAS číslo: 54-20-6 EINECS: 200-197-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...