Tisk…

4-Amino-7-(trifluoromethyl)quinoline

Dodavatel: Apollo Scientific
Varování

PC7756-1G PC7756-250MG PC7756-500MG
APOSPC7756-1GEA 5790 CZK
APOSPC7756-1G APOSPC7756-250MG APOSPC7756-500MG
4-Amino-7-(trifluoromethyl)quinoline
4-Amino-7-(trifluoromethyl)quinoline
Vzorec: C₁₀H₇F₃N₂
MW: 212.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269411
CAS číslo: 243666-11-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR