Tisk…

(2-Bromo-5-fluorophenyl)acetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Bromo-5-fluorophenylacetic acid

PC7747-25G PC7747-100G
APOSPC7747-25GEA 2610 CZK
APOSPC7747-25G APOSPC7747-100G
(2-Bromo-5-fluorophenyl)acetic acid 98%
(2-Bromo-5-fluorophenyl)acetic acid
Vzorec: C₈H₆BrFO₂
MW: 233,03 g/mol
Bod tání: 113…114 °C
Storage Temperature: Ambient
MDL Number: MFCD03094440
CAS číslo: 739336-26-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR