Tisk…

5-Fluoro-2-methoxybenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (5-Fluoro-2-methoxyphenyl)methanol

PC7735-1G PC7735-5G PC7735-10G
APOSPC7735-1GEA 622 CZK
APOSPC7735-1G APOSPC7735-5G APOSPC7735-10G
5-Fluoro-2-methoxybenzyl alcohol
5-Fluoro-2-methoxybenzyl alcohol
Vzorec: C₈H₉FO₂
MW: 156,16 g/mol
Bod tání: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00671767
CAS číslo: 426831-32-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR