2-(Trifluoromethyl)-DL-phenylalanine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Amino-3-(2-(trifluoromethyl)phenyl)propanoic acid
PC7713X-1G
PC7713X-5G
PC7713X-25G
APOSPC7713X-1GEA
1740
CZK
APOSPC7713X-1G
APOSPC7713X-5G
APOSPC7713X-25G
2-(Trifluoromethyl)-DL-phenylalanine
2-(Trifluoromethyl)-DL-phenylalanine
Vzorec:
C₁₀H₁₀F₃NO₂ MW: 233,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061305 CAS číslo: 3832-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...