Tisk…

3-Benzyloxy-6-bromo-2-fluorophenylboronic acid 98%

Dodavatel: Apollo Scientific

PC7705-1G PC7705-5G
APOSPC7705-1GEA 3940 CZK
APOSPC7705-1G APOSPC7705-5G
3-Benzyloxy-6-bromo-2-fluorophenylboronic acid 98%
3-Benzyloxy-6-bromo-2-fluorophenylboronic acid
Vzorec: C₁₃H₁₁BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD09475868
CAS číslo: 957035-10-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR