Tisk…

2',4'-Difluoro-3'-(trifluoromethyl)acetophenone 98%

Dodavatel: Apollo Scientific
Varování

PC7673-1G PC7673-250MG PC7673-5G
APOSPC7673-1GEA 5720 CZK
APOSPC7673-1G APOSPC7673-250MG APOSPC7673-5G
2',4'-Difluoro-3'-(trifluoromethyl)acetophenone 98%
2',4'-Difluoro-3'-(trifluoromethyl)acetophenone
Vzorec: C₉H₅F₅O
MW: 224,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547946
CAS číslo: 1202679-46-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR