Tisk…

1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC7666-1G PC7666-100MG
APOSPC7666-1GEA 3310 CZK
APOSPC7666-1G APOSPC7666-100MG
1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulphonyl chloride
1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulphonyl chloride
Vzorec: C₉H₅ClF₃N₃O₂S
Bod tání: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD08689729
CAS číslo: 1006441-36-6
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR